CH6371382757 Chem Impex International, Inc.

4-Chloroindoline

  • MDL number MFCD07371635
  • Cas Number : 41910-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-2,3-dihydro-1H-indole
Purity 97
Storage temp. Store at 0-8
Physical Form Colorless to light yellow clear liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-chloroindoline/8778?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-chloroindoline/8778?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart