CH6371382758 Chem Impex International, Inc.

4-Chloro-2-anisidine hydrochloride

  • MDL number MFCD00068437
  • Cas Number : 93-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-5-chloroanisole hydrochloride
Purity 95
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-chloro-2-anisidine-hydrochloride/8779?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-chloro-2-anisidine-hydrochloride/8779?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart