CH6371382764 Chem Impex International, Inc.

4-Methoxy-1H-indole-3-carboxylic acid

  • MDL number MFCD05664011
  • Cas Number : 203937-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Tan solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methoxy-1-i-h-i-indole-3-carboxylic-acid/8785?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methoxy-1-i-h-i-indole-3-carboxylic-acid/8785?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart