CH6371382769 Chem Impex International, Inc.

2-Methoxy-5-(1,3-dioxoisoindolin-2-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD03453269
  • Cas Number : 885279-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-methoxy-5-1-3-dioxoisoindolin-2-yl-benzene-1-sulfonyl-chloride/8792?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-methoxy-5-1-3-dioxoisoindolin-2-yl-benzene-1-sulfonyl-chloride/8792?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart