CH6371382771 Chem Impex International, Inc.

5-(3-Butyl ureido)-2-ethoxy benzenesulfonyl chloride

  • MDL number MFCD03424977
  • Cas Number : 680618-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Yellow oily liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-3-butyl-ureido-2-ethoxy-benzenesulfonyl-chloride/8797?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-3-butyl-ureido-2-ethoxy-benzenesulfonyl-chloride/8797?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart