CH6371382773 Chem Impex International, Inc.

3-(3-Chlorophenyl)-1-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD06738764
  • Cas Number : 1006463-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-3-chlorophenyl-1-phenyl-1-i-h-i-pyrazol-5-amine/8799?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-3-chlorophenyl-1-phenyl-1-i-h-i-pyrazol-5-amine/8799?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart