CH6371382779 Chem Impex International, Inc.

5-(4-Chlorophenyl)-2H-tetrazole

  • MDL number MFCD00172856
  • Cas Number : 16687-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256-262 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystals
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-4-chlorophenyl-2-i-h-i-tetrazole/8806?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-chlorophenyl-2-i-h-i-tetrazole/8806?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart