CH6371382786 Chem Impex International, Inc.

5-Ethyl-2-methoxy-benzenesulfonyl chloride

  • MDL number MFCD03424997
  • Cas Number : 88040-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60 - 66 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-ethyl-2-methoxy-benzenesulfonyl-chloride/8815?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-ethyl-2-methoxy-benzenesulfonyl-chloride/8815?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart