CH6371382795 Chem Impex International, Inc.

6-Amino-pyridin-3-ol hydrochloride

  • MDL number MFCD14586323
  • Cas Number : 55717-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Light brown solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-amino-pyridin-3-ol-hydrobromide/8828?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-amino-pyridin-3-ol-hydrobromide/8828?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart