CH6371382857 Chem Impex International, Inc.

(4-Bromo-3-methylphenyl)hydrazine

  • MDL number MFCD02663108
  • Cas Number : 112626-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-bromo-3-methylphenyl-hydrazine/8908?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-bromo-3-methylphenyl-hydrazine/8908?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart