CH6371382867 Chem Impex International, Inc.

(R)-4-Boc-thiomorpholine-3-carboxylic acid

  • MDL number MFCD04114906
  • Cas Number : 114525-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (R)-4-(tert-Butoxycarbonyl)-thiomorpholine-3-carboxylic acid
Purity 97
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-4-boc-thiomorpholine-3-carboxylic-acid/8926?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-4-boc-thiomorpholine-3-carboxylic-acid/8926?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart