CH6371382868 Chem Impex International, Inc.

(R)-Thiomorpholine-3-carboxylic acid

  • MDL number MFCD01318713
  • Cas Number : 65527-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-thiomorpholine-3-carboxylic-acid/8928?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-thiomorpholine-3-carboxylic-acid/8928?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart