CH6371382907 Chem Impex International, Inc.

1H-Indole-7-carbohydrazide

  • MDL number MFCD00572868
  • Cas Number : 321309-24-4
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-i-h-i-indole-7-carbohydrazide/8996?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-i-h-i-indole-7-carbohydrazide/8996?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart