CH6371382919 Chem Impex International, Inc.

2-(3-Bromophenyl)thiazole-4-carbaldehyde

  • MDL number MFCD06337054
  • Cas Number : 750624-69-2
  • Molecular Weight : 268.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8°C
Physical Form Pale white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-3-bromophenyl-thiazole-4-carbaldehyde/9012?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-3-bromophenyl-thiazole-4-carbaldehyde/9012?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart