CH6371382923 Chem Impex International, Inc.

2-(3-Methoxyphenyl)thiazole-4-carbaldehyde

  • MDL number MFCD06335753
  • Cas Number : 749902-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form Off-white to yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-3-methoxyphenyl-thiazole-4-carbaldehyde/9018?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-3-methoxyphenyl-thiazole-4-carbaldehyde/9018?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart