CH6371382924 Chem Impex International, Inc.

2-(3-Methoxyphenyl)thiazole-4-carboxylic acid ethyl ester

  • MDL number MFCD06738341
  • Cas Number : 115299-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form Pale yellow liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-3-methoxyphenyl-thiazole-4-carboxylic-acid-ethyl-ester/9019?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-3-methoxyphenyl-thiazole-4-carboxylic-acid-ethyl-ester/9019?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart