CH6371382926 Chem Impex International, Inc.

2-[(4-Bromophenoxy)methyl]oxirane

  • MDL number MFCD00022341
  • Cas Number : 2212-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-52 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-bromophenoxy-methyl-oxirane/9021?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-bromophenoxy-methyl-oxirane/9021?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart