CH6371382979 Chem Impex International, Inc.

3-Amino-tetrahydro-thiophene-3-carboxylic acid

  • MDL number MFCD01318281
  • Cas Number : 32418-99-8
  • Molecular Weight : 147.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-amino-tetrahydro-thiophene-3-carboxylic-acid/9097?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-amino-tetrahydro-thiophene-3-carboxylic-acid/9097?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart