CH6371382981 Chem Impex International, Inc.

3-Bromo-4-methoxy-pyridine

  • MDL number MFCD05664024
  • Cas Number : 82257-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form Brown Liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-bromo-4-methoxy-pyridine/9101?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-bromo-4-methoxy-pyridine/9101?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart