CH6371382988 Chem Impex International, Inc.

tert-Butyl 3-nitrobenzoate

  • MDL number MFCD02091504
  • Cas Number : 58656-99-8
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form Brown solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-tert-i-butyl-3-nitrobenzoate/9111?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-tert-i-butyl-3-nitrobenzoate/9111?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart