CH6371383004 Chem Impex International, Inc.

4-Benzyl-2-[(piperidin-1-yl)methyl]morpholine

  • MDL number MFCD06658346
  • Cas Number : 874881-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form Orange liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-benzyl-2-piperidin-1-yl-methyl-morpholine/9134?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-benzyl-2-piperidin-1-yl-methyl-morpholine/9134?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart