CH6371383019 Chem Impex International, Inc.

4-Phenyl-pyridin-2-ylamine

  • MDL number MFCD00137848
  • Cas Number : 60781-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-4-phenylpyridine
Purity 97
Storage temp. Store at 0-8
Physical Form Yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-phenyl-pyridin-2-ylamine/9155?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-phenyl-pyridin-2-ylamine/9155?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart