CH6371383026 Chem Impex International, Inc.

5-(4-Bromophenyl)-2-phenyl-2H-pyrazol-3-ylamine

  • MDL number MFCD00197060
  • Cas Number : 201735-04-8
  • Molecular Weight : 314.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209 - 215 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form Pale yellow flakes
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-4-bromophenyl-2-phenyl-2h-pyrazol-3-ylamine/9166?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-bromophenyl-2-phenyl-2h-pyrazol-3-ylamine/9166?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart