CH6371383040 Chem Impex International, Inc.

5-Cyano-1H-indole-2-boronic acid

  • MDL number MFCD06798223
  • Cas Number : 871329-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-cyano-1-i-h-i-indole-2-boronic-acid/9184?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-cyano-1-i-h-i-indole-2-boronic-acid/9184?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart