CH6371383066 Chem Impex International, Inc.

6-Phenyl-pyridin-2-ylamine

  • MDL number MFCD00234719
  • Cas Number : 39774-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Amino-2-phenylpyridine
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-phenyl-pyridin-2-ylamine/9221?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-phenyl-pyridin-2-ylamine/9221?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart