CH6371383067 Chem Impex International, Inc.

6-Propyl-pyridin-2-ylamine

  • MDL number MFCD00130076
  • Cas Number : 41995-29-3
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Brown liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/6-propyl-pyridin-2-ylamine/9222?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/6-propyl-pyridin-2-ylamine/9222?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart