CH6371383102 Chem Impex International, Inc.

Dimethyl-(4-pyrrolidin-2-yl-phenyl)amine

  • MDL number MFCD02663403
  • Cas Number : 298690-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dimethyl-4-pyrrolidin-2-yl-phenyl-amine/9262?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dimethyl-4-pyrrolidin-2-yl-phenyl-amine/9262?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart