CH6371383117 Chem Impex International, Inc.

Z-piperazine-2-carboxylic acid

  • MDL number MFCD04972466
  • Cas Number : 129365-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-piperazine-2-carboxylic-acid/9291?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-piperazine-2-carboxylic-acid/9291?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart