CH6371383135 Chem Impex International, Inc.

(2-Fluorophenyl)oxo-acetonitrile

  • MDL number MFCD06658449
  • Cas Number : 80277-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluorobenzoyl cyanide
Purity 95
Storage temp. Store at 0-8
Physical Form Yellow liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-fluorophenyl-oxo-acetonitrile/9321?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-fluorophenyl-oxo-acetonitrile/9321?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart