CH6371383147 Chem Impex International, Inc.

(4-Benzyl-morpholin-2-ylmethyl)dimethylamine

  • MDL number MFCD06658340
  • Cas Number : 122894-38-6
  • Molecular Weight : 234.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form Yellow liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-benzyl-morpholin-2-ylmethyl-dimethylamine/9361?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-benzyl-morpholin-2-ylmethyl-dimethylamine/9361?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart