CH6371383154 Chem Impex International, Inc.

(4-Ethoxyphenyl)oxo-acetonitrile

  • MDL number MFCD06658436
  • Cas Number : 62869-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-ethoxyphenyl-oxo-acetonitrile/9369?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-ethoxyphenyl-oxo-acetonitrile/9369?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart