CH6371383155 Chem Impex International, Inc.

(4-Fluoro-3-nitrobenzyl)carbamic acid tert-butyl ester

  • MDL number MFCD06658388
  • Cas Number : 885280-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-fluoro-3-nitrobenzyl-carbamic-acid-i-tert-i-butyl-ester/9370?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-fluoro-3-nitrobenzyl-carbamic-acid-i-tert-i-butyl-ester/9370?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart