CH6371383174 Chem Impex International, Inc.

6-Acetyl-5-hydroxytetralin

  • MDL number MFCD06658289
  • Cas Number : 95517-07-0
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/6-acetyl-5-hydroxytetralin

Safety Information

MSDS http://www.chemimpex.com/6-acetyl-5-hydroxytetralin
Availability details for
Quantity
  • Availability
  • Add to Cart