CH6371383181 Chem Impex International, Inc.

1-(4-Chloropyridin-2-yl)piperazine

  • MDL number MFCD06738950
  • Cas Number : 885277-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-4-chloropyridin-2-yl-piperazine-trihydrochloride/9494?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-4-chloropyridin-2-yl-piperazine-trihydrochloride/9494?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart