CH6371383184 Chem Impex International, Inc.

1-(4-Hydroxy-7-methyl-indan-5-yl)ethanone

  • MDL number MFCD00173684
  • Cas Number : 175136-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-4-hydroxy-7-methyl-indan-5-yl-ethanone/9499?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-4-hydroxy-7-methyl-indan-5-yl-ethanone/9499?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart