CH6371383191 Chem Impex International, Inc.

1-(4-(Trifluoromethoxy)phenyl)piperazin-2-one hydrochloride

  • MDL number MFCD06658518
  • Cas Number : 267659-71-2
  • Molecular Weight : 296.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-4-trifluoromethoxy-phenyl-piperazin-2-one-hydrochloride/9509?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-4-trifluoromethoxy-phenyl-piperazin-2-one-hydrochloride/9509?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart