CH6371383199 Chem Impex International, Inc.

1,5-Dihydro-pyrrolo[3,2-d]pyrimidine-2,4-dione

  • MDL number MFCD08059275

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1H-Pyrrolo[3,2-d]pyrimidine-2,4(3H,5H)-dione
Purity 0.96
Storage temp. Store at 0-8
Physical Form Yellow solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-5-dihydro-pyrrolo-3-2-i-d-i-pyrimidine-2-4-dione/9523?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-5-dihydro-pyrrolo-3-2-i-d-i-pyrimidine-2-4-dione/9523?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart