CH6371383220 Chem Impex International, Inc.

1-(1-(4-Chlorophenyl)vinyl)benzene

  • MDL number MFCD06658410
  • Cas Number : 18218-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form Light yellow liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-1-4-chlorophenyl-vinyl-benzene/9562?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-1-4-chlorophenyl-vinyl-benzene/9562?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart