CH6371383231 Chem Impex International, Inc.

1H-Indole-4-carboxylic acid hydrazide

  • MDL number MFCD06738885
  • Cas Number : 885272-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Off-white to pale yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1h-indole-4-carboxylic-acid-hydrazide/9585?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1h-indole-4-carboxylic-acid-hydrazide/9585?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart