CH6371383247 Chem Impex International, Inc.

2-(2,3-Dihydro-benzo[1,4]dioxin-2-yl)ethylamine

  • MDL number MFCD01659461
  • Cas Number : 87086-36-0
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Colorless liquid
Shipping Temperature RT
Country of Origin USA
Boiling point 113-120 °C / 3 mm
Product Link http://www.chemimpex.com/product/productinfo/2-2-3-dihydro-benzo-1-4-dioxin-2-yl-ethylamine/9610?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-2-3-dihydro-benzo-1-4-dioxin-2-yl-ethylamine/9610?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart