CH6371383249 Chem Impex International, Inc.

2-(2,6-Dichlorophenyl)succinic acid

  • MDL number MFCD06658425
  • Cas Number : 42474-07-7
  • Molecular Weight : 263.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-2-6-dichlorophenyl-succinic-acid/9612?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-2-6-dichlorophenyl-succinic-acid/9612?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart