CH6371383252 Chem Impex International, Inc.

2-(2-Bromophenyl)succinic acid

  • MDL number MFCD06658382
  • Cas Number : 20608-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-2-bromophenyl-succinic-acid/9621?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-2-bromophenyl-succinic-acid/9621?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart