CH6371383260 Chem Impex International, Inc.

2-(3,4-Dichlorophenyl)succinic acid

  • MDL number MFCD06658426
  • Cas Number : 93553-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-3-4-dichlorophenyl-succinic-acid/9645?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-3-4-dichlorophenyl-succinic-acid/9645?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart