CH6371383266 Chem Impex International, Inc.

2-(3-Methoxybenzyl)-3-oxo-hexanoic acid ethyl ester

  • MDL number MFCD06658445
  • Cas Number : 885279-96-9
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8
Physical Form Light yellow liquid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/2-3-methoxybenzyl-3-oxo-hexanoic-acid-ethyl-ester/9675?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-3-methoxybenzyl-3-oxo-hexanoic-acid-ethyl-ester/9675?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart