CH6371383269 Chem Impex International, Inc.

2-(4-Benzyloxyphenyl)thiazole-4-carboxylic acid ethyl ester

  • MDL number MFCD06738333
  • Cas Number : 885279-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Pale yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-benzyloxyphenyl-thiazole-4-carboxylic-acid-ethyl-ester/9688?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-benzyloxyphenyl-thiazole-4-carboxylic-acid-ethyl-ester/9688?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart