CH6371383271 Chem Impex International, Inc.

2-(4-Bromophenyl)succinic acid

  • MDL number MFCD06656575
  • Cas Number : 71471-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-bromophenyl-succinic-acid/9697?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-bromophenyl-succinic-acid/9697?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart