CH6371383277 Chem Impex International, Inc.

2-(4-Methoxyphenyl)succinic acid

  • MDL number MFCD00778506
  • Cas Number : 6331-59-5
  • Molecular Weight : 224.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white to white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-methoxyphenyl-succinic-acid/9711?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-methoxyphenyl-succinic-acid/9711?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart