CH6371383280 Chem Impex International, Inc.

2-(O-Tolyl)thiazole-4-carboxylic acid ethyl ester

  • MDL number MFCD06738344
  • Cas Number : 885278-51-3
  • Molecular Weight : 247.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Yellow liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-o-tolyl-thiazole-4-carboxylic-acid-ethyl-ester/9729?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-o-tolyl-thiazole-4-carboxylic-acid-ethyl-ester/9729?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart