CH6371383301 Chem Impex International, Inc.

2-Bromo-3-nitro-benzoic acid ethyl ester

  • MDL number MFCD00024177
  • Cas Number : 31706-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Light yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-bromo-3-nitro-benzoic-acid-ethyl-ester/9771?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-bromo-3-nitro-benzoic-acid-ethyl-ester/9771?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart