CH6371383302 Chem Impex International, Inc.

2-Chloro-3-methoxy-5-nitro-pyridine

  • MDL number MFCD06658397
  • Cas Number : 75711-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-chloro-3-methoxy-5-nitro-pyridine/9772?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-chloro-3-methoxy-5-nitro-pyridine/9772?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart